2,2,6,6-四甲基哌啶 98%
供应商: Thermo Fisher Scientific
|
Danger
|
151950100
151950250
ACRO151950100EA
0
CNY
ACRO151950100
ACRO151950250
2,2,6,6-四甲基哌啶 98%
2,2,6,6-四甲基哌啶
分子式:
C₉H₁₉N 分子量: 141.26 g/mol 沸点: 152 °C (760 mmHg) 密度: 0.83 g/cm³ 闪点: 24 °C 存储条件: Ambient |
MDL编号:
MFCD00005985 CAS号: 768-66-1 EINECS: 212-199-3 UN号: 1992 ADR: 3 III |
产品技术规格
Appearance | Clear colorless to yellow to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4425 to 1.4465 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...