2-吡咯烷酮 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
丁内酰胺, 2-吡咯酮
132112500
132115000
ACRO132112500EA
0
CNY
ACRO132112500
ACRO132115000
2-吡咯烷酮 99%
2-吡咯烷酮
分子式:
C₄H₇NO 沸点: 250 °C (760 mmHg) 密度: 1,103 g/cm³ 闪点: 138 °C 存储条件: Ambient |
MDL编号:
MFCD00005270 CAS号: 616-45-5 EINECS: 210-483-1 Merck Index: 12,08200 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid or low melting mass (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.4860 to 1.4880 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...