4-戊烯-1-醇 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 2-烯丙基乙醇

150010100 150010500
ACRO150010100EA 0 CNY
ACRO150010100 ACRO150010500
4-戊烯-1-醇 99%
4-戊烯-1-醇
分子式: C₅H₁₀O
分子量: 86.13 g/mol
沸点: 137
密度: 0.834 g/cm³
闪点: 43 °C
存储条件: Ambient
MDL编号: MFCD00002975
CAS号: 821-09-0
EINECS: 212-473-2
UN号: 1987

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4289 to 1.4309 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR