马来酸二乙酯 97%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
顺丁烯二酸二乙酯
114440010
114440050
114442500
ACRO114440010EA
0
CNY
ACRO114440010
ACRO114440050
ACRO114442500
马来酸二乙酯 97%
马来酸二乙酯
分子式:
C₈H₁₂O₄ 分子量: 172.18 g/mol 沸点: 225 溶点: –10 °C 密度: 1.06 g/cm³ 闪点: 93 存储条件: Ambient |
MDL编号:
MFCD00009191 CAS号: 141-05-9 EINECS: 205-451-9 Merck Index: 13,03150 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4380 to 1.4420 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...