顺丁烯二酸二丁酯 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: Maleic acid dibutyl ester

353100010 353102500
ACRO353100010EA 0 CNY
ACRO353100010 ACRO353102500
顺丁烯二酸二丁酯 97%
顺丁烯二酸二丁酯
分子式: C₁₂H₂₀O₄
分子量: 228.29 g/mol
沸点: 274…277 °C
密度: 0.99 g/cm³
闪点: 120 °C
存储条件: Ambient
MDL编号: MFCD00009447
CAS号: 105-76-0
EINECS: 203-328-4
UN号: 3082
ADR: 9 III

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR