3,4-二甲氧基苯乙酸 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
3,4-二甲氧基苯乙酸, 高藜芦酸
115611000
115615000
115610250
ACRO115611000EA
0
CNY
ACRO115611000
ACRO115615000
ACRO115610250
3,4-二甲氧基苯乙酸 99%
3,4-二甲氧基苯乙酸
分子式:
(C₆H₃(OCH₃)₂)CH₂COOH 分子量: 196.2 g/mol 沸点: 327 溶点: 96…99 °C 存储条件: Ambient |
MDL编号:
MFCD00004335 CAS号: 93-40-3 EINECS: 202-244-5 |
产品技术规格
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Melting point | 96 to 99 °C |
Water | ≤1 % (K.F.) |
Titration with TBAH | ≥98.5 % (On dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...