Homovanillic acid 98%
供应商: Thermo Fisher Scientific
|
Warning
|
162530010
162530050
ACRO162530010EA
0
CNY
ACRO162530010
ACRO162530050
Homovanillic acid 98%
Homovanillic acid
分子式:
(C₆H₃(OH)(OCH₃))CH₂COOH 沸点: 339 °C (1013 hPa) 溶点: 141…145 °C |
MDL编号:
MFCD00004350 CAS号: 306-08-1 EINECS: 206-176-7 Merck Index: 13,04761 |
产品技术规格
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 141 to 145 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...