2-甲基-1-丙醇 99+%, Spectrophotometric Grade
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
异丁醇, 异丙基甲醇
167700010
167705000
ACRO167700010EA
0
CNY
ACRO167700010
ACRO167705000
2-甲基-1-丙醇 99+%, Spectrophotometric Grade
2-甲基-1-丙醇
分子式:
(CH₃)₂CHCH₂OH 分子量: 74.12 g/mol 沸点: 107.9 溶点: –108 °C 密度: 0.8 g/cm³ 闪点: 28 存储条件: Ambient |
MDL编号:
MFCD00004740 CAS号: 78-83-1 EINECS: 201-148-0 UN号: 1212 ADR: 3 III Merck Index: 13,05148 |
|
产品技术规格
Appearance | Clear liquid |
Water | ≤0.05 % |
GC | ≥99.0 % |
UV | at 330 nm A: ≤0.01 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...