异丁酸 99+%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-甲基丙酸
122520010
122520250
122525000
ACRO122520010EA
0
CNY
ACRO122520010
ACRO122520250
ACRO122525000
异丁酸 99+%
异丁酸
分子式:
C₄H₈O₂ 沸点: 155 °C (1013 hPa) 溶点: –47 °C 密度: 0,95 g/cm³ 闪点: 59 °C 存储条件: Ambient |
MDL编号:
MFCD00002658 CAS号: 79-31-2 EINECS: 201-195-7 UN号: 2529 ADR: 3,III Merck Index: 13,05172 |
产品技术规格
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3920 to 1.3940 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...