J.T.Baker® BAKERBOND Speedisk® Hydrophobic DVB, SPE Columns
供应商: Avantor
BAKR8109-06EA 2226 CNY
BAKR8109-06 BAKR8109-09
J.T.Baker® BAKERBOND Speedisk® Hydrophobic DVB, SPE Columns
色谱柱
These columns feature BAKERBOND® DVB polymer resins, which are the product of ultra-clean polymer microparticle technology. These resin particles have a large surface area, are highly rigid, are stable over pH range 1 to 14, are water-wettable and are not impacted by sorbent drying. These columns are recommended when advanced detection methods are used.
  • Improve sample preparation recovery
  • Polymer based
General applications: Slightly polar to non polar analytes.
These SPE columns can be used in adsorption mode.