甲烷磺酸银 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 甲烷磺酸

419340250 419340050
ACRO419340250EA 0 CNY
ACRO419340250 ACRO419340050
甲烷磺酸银 99%
甲烷磺酸银
分子式: CH₃AgO₃S
分子量: 202.97 g/mol
溶点: 252…256 °C
存储条件: Ambient
MDL编号: MFCD00064795
CAS号: 2386-52-9
EINECS: 219-199-2

现在订购

产品技术规格

Appearance White to beige to light grey Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR