叔丁醇钾 98+%, pure
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
第三丁氧基钾, 次丁氧基钾
168880050
168881000
168885000
ACRO168880050EA
0
CNY
ACRO168880050
ACRO168881000
ACRO168885000
叔丁醇钾 98+%, pure
叔丁醇钾
分子式:
C₄H₉KO 溶点: 240…258 °C 存储条件: Ambient |
MDL编号:
MFCD00012162 CAS号: 865-47-4 EINECS: 212-740-3 UN号: 3206 ADR: 4.2,II |
产品技术规格
Appearance | White to off-white Powder |
Titration with HCl | ≥98.0 % |
KOH + K2CO3 (as KOH) | ≤0.50 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...