乙酸酐 97+% ACS
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Acetyl oxide, Acetyl acetate
423230010
423235000
ACRO423230010EA
0
CNY
ACRO423230010
ACRO423235000
乙酸酐 97+% ACS
乙酸酐
分子式:
(CH₃CO)₂O 沸点: 139,9 °C (1013 hPa) 密度: 1,087 g/cm³ 闪点: 54 °C 存储条件: Ambient |
MDL编号:
MFCD00008705 CAS号: 108-24-7 EINECS: 203-564-8 UN号: 1715 ADR: 8,II REACH: 01-2119486470-36 Merck Index: 13,00057 |
|
产品技术规格
Appearance | Clear liquid |
GC | ≥97.0 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.003 % |
Chloride (Cl) | ≤5 ppm |
Phosphate (PO4) | ≤10 ppm |
Sulfate (SO4) | ≤5 ppm |
Iron (Fe) | ≤5 ppm |
Heavy metals (as Pb) | ≤2 ppm |
KMnO4-reducing substance | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...