二乙二醇二乙醚 98+%, extra pure

供应商: Thermo Fisher Scientific
Warning

Synonyms: 二乙基卡必醇, 2-乙氧基乙基醚, 双(2-乙氧基乙基)醚

117900010 117900025 117905000
ACRO117900010EA 0 CNY
ACRO117900010 ACRO117900025 ACRO117905000
二乙二醇二乙醚 98+%, extra pure
二乙二醇二乙醚
分子式: C₈H₁₈O₃
分子量: 162.23 g/mol
沸点: 180…190 °C (760 mmHg)
溶点: –44.3 °C
密度: 0.9 g/cm³
闪点: 82 °C
存储条件: Ambient
MDL编号: MFCD00009254
CAS号: 112-36-7
EINECS: 203-963-7
Merck Index: 13,03145

现在订购

产品技术规格

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4110 to 1.4130 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥98.0 %
Color scale ≤15 APHA
Residue after evaporation ≤0.05 %
Acidity (CH3COOH) ≤0.015 %
Peroxides ≤0.0015 % (as H2O2)
Iron (Fe) ≤3 ppm
Magnesium (Mg) ≤0.002 %
Sodium (Na) ≤0.005 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR