原丙酸三乙酯 98+%

供应商: Thermo Fisher Scientific
Warning

139671000 139675000
ACRO139671000EA 0 CNY
ACRO139671000 ACRO139675000
原丙酸三乙酯 98+%
原丙酸三乙酯
分子式: C₉H₂₀O₃
沸点: 155…160 °C (760 mmHg)
溶点: < 0 °C
密度: 0,886 g/cm³
闪点: 60 °C
存储条件: Ambient
MDL编号: MFCD00009226
CAS号: 115-80-0
EINECS: 204-108-0
UN号: 3271
ADR: 3,III

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4010 to 1.4030 (20°C, 589 nm)
GC ≥98.0 %
Specific optical rotation +18° to +28° (20°C, 589 nm) (c=2,ethanol/0.5N HCl(4:1))


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR