二乙胺 99+%, 超纯

供应商: Thermo Fisher Scientific
Danger

149450010 149450025
ACRO149450010EA 0 CNY
ACRO149450010 ACRO149450025
二乙胺 99+%, 超纯
二乙胺
分子式: (C₂H₅)₂NH
沸点: 55 °C (760 mmHg)
溶点: –50 °C
密度: 0,71 g/cm³
闪点: –23 °C
存储条件: Ambient
MDL编号: MFCD00009032
CAS号: 109-89-7
EINECS: 203-716-3
UN号: 1154
ADR: 3,II
Merck Index: 13,03138
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c054fa528239af221505ef08b7c0d2503c9d1d7&vid=e1e59ef071f0c580f9eb9716f4801d314ff824c3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3840 to 1.3860 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR