2-(2-氨基乙氧基)乙醇 98%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
二甘醇胺
103630010
103631000
ACRO103630010EA
0
CNY
ACRO103630010
ACRO103631000
2-(2-氨基乙氧基)乙醇 98%
2-(2-氨基乙氧基)乙醇
分子式:
C₄H₁₁NO₂ 分子量: 105.14 g/mol 沸点: 218…224 °C (760 mmHg) 溶点: –12.5 °C 密度: 1.06 g/cm³ 闪点: 127 °C 存储条件: Ambient |
MDL编号:
MFCD00008181 CAS号: 929-06-6 EINECS: 213-195-4 UN号: 3055 ADR: 8 III |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4570 to 1.4610 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...