碘乙酸 97%
供应商: Thermo Fisher Scientific
|
Danger
|
122280250
122280500
122281000
ACRO122280250EA
0
CNY
ACRO122280250
ACRO122280500
ACRO122281000
碘乙酸 97%
碘乙酸
分子式:
C₂H₃IO₂ 沸点: 262 °C (1013 hPa) 溶点: 79…83 °C 密度: 2,2694 g/cm³ (85 °C) 存储条件: Refrigerator |
MDL编号:
MFCD00002685 CAS号: 64-69-7 EINECS: 200-590-1 UN号: 2923 ADR: 8,I Merck Index: 13,05047 |
产品技术规格
Appearance | Yellow to orange-brown Powder or crystals or flakes |
Infrared spectrum | Conforms |
Melting point | 76 to 83 °C |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...