七氟丁酸 99%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
HFBA, 全氟丁酸, Edman 试剂3
172800250
172801000
ACRO172800250EA
0
CNY
ACRO172800250
ACRO172801000
七氟丁酸 99%
七氟丁酸
分子式:
C₄HF₇O₂ 沸点: 120 °C (755 mmHg) 密度: 1,64 g/cm³ 存储条件: Ambient |
MDL编号:
MFCD00004171 CAS号: 375-22-4 EINECS: 206-786-3 UN号: 3265 ADR: 8,I |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Titration with NaOH | ≥98.5 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...