丁二酸二甲酯 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
琥珀酸二甲酯, 丁二酸二甲酯
150731000
150735000
ACRO150731000EA
0
CNY
ACRO150731000
ACRO150735000
丁二酸二甲酯 99%
丁二酸二甲酯
分子式:
C₆H₁₀O₄ 沸点: 200 °C (1013 hPa) 溶点: 18…19 °C 密度: 1,11 g/cm³ 闪点: 85 °C 存储条件: Ambient |
MDL编号:
MFCD00008466 CAS号: 106-65-0 EINECS: 203-419-9 Merck Index: 12,09038 |
产品技术规格
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4180 to 1.4210 (20°C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤25 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...