肉豆蔻酸甲酯 99%
供应商: Thermo Fisher Scientific
Synonyms:
Myristic acid methyl ester, 十四酸甲酯
165010025
165011000
165015000
ACRO165010025EA
0
CNY
ACRO165010025
ACRO165011000
ACRO165015000
肉豆蔻酸甲酯 99%
肉豆蔻酸甲酯
分子式:
C₁₅H₃₀O₂ 分子量: 242.4 g/mol 沸点: 323 溶点: 18.4 °C 密度: 0.863 g/cm³ 闪点: >110 °C 存储条件: Refrigerator |
MDL编号:
MFCD00008983 CAS号: 124-10-7 EINECS: 204-680-1 |
产品技术规格
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4340 to 1.4380 (20°C, 589 nm) |
Density | (20/4°C) 0.853 to 0.873 g/ml |
GC | ≥98.5 % |
Acid value | ≤1 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...