2-乙酰胺基苯酚 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 2′-羟基乙酰苯胺

102310250 102311000
ACRO102310250EA 0 CNY
ACRO102310250 ACRO102311000
2-乙酰胺基苯酚 97%
2-乙酰胺基苯酚
分子式: C₈H₉NO₂
分子量: 151.16 g/mol
溶点: 207…210 °C
MDL编号: MFCD00002181
CAS号: 614-80-2
EINECS: 210-396-9

现在订购

产品技术规格

Appearance Off-white to beige to brown Powder
Infrared spectrum Conforms
Melting point 205 to 210 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR