丁酰氯 99%
供应商: Thermo Fisher Scientific
|
Danger
|
149210010
149212500
ACRO149210010EA
0
CNY
ACRO149210010
ACRO149212500
丁酰氯 99%
丁酰氯
分子式:
C₄H₇ClO 分子量: 106.55 g/mol 沸点: 102 °C (760 mmHg) 溶点: –89 °C 密度: 1.018 g/cm³ 闪点: 18 °C 存储条件: Ambient |
MDL编号:
MFCD00000752 CAS号: 141-75-3 EINECS: 205-498-5 UN号: 2353 ADR: 3 II Merck Index: 13,01598 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4112 to 1.4132 (20°C, 589 nm) |
Titration RCOCl | ≥98.5 % |
Titration RCOOH | ≤2 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...