癸二腈 98%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1,8-亚辛基二氰化物, Decanedinitrile
419250250
419251000
ACRO419250250EA
0
CNY
ACRO419250250
ACRO419251000
癸二腈 98%
癸二腈
分子式:
C₁₀H₁₆N₂ 分子量: 164.25 g/mol 沸点: 332 溶点: 7 °C 密度: 0.91 g/cm³ 闪点: >110 °C |
MDL编号:
MFCD00019900 CAS号: 1871-96-1 EINECS: 217-494-0 UN号: 3276 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4470 to 1.4490 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...