对氯乙酰苯胺 97%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-氯乙酰苯胺
169950050
169950500
ACRO169950050EA
0
CNY
ACRO169950050
ACRO169950500
对氯乙酰苯胺 97%
对氯乙酰苯胺
分子式:
C₈H₈ClNO 分子量: 169.61 g/mol 沸点: 333 °C (760 mmHg) 溶点: 177…181 °C 存储条件: Ambient |
MDL编号:
MFCD00000612 CAS号: 539-03-7 EINECS: 208-707-8 Merck Index: 13,02128 |
产品技术规格
Appearance | White to grey-beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 177 to 181 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...