5-溴戊酸乙酯 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 5-Bromovaleric acid ethyl ester, Ethyl 5-bromovalerate

156540250 156541000
ACRO156540250EA 0 CNY
ACRO156540250 ACRO156541000
5-溴戊酸乙酯 99%
5-溴戊酸乙酯
分子式: BrH₂C(CH₂)₃COOCH₂CH₃
沸点: 104…109 °C (12 mmHg)
密度: 1,32 g/cm³
闪点: 103 °C
MDL编号: MFCD00000266
CAS号: 14660-52-7
EINECS: 238-705-2

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4570 to 1.4600 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR