溴乙酰溴 98%

供应商: Thermo Fisher Scientific
Danger

106601000 106605000
ACRO106601000EA 0 CNY
ACRO106601000 ACRO106605000
溴乙酰溴 98%
溴乙酰溴
分子式: BrCH₂COOBr
分子量: 201.85 g/mol
沸点: 148
密度: 2.31 g/cm³
闪点: >105 °C
存储条件: Refrigerator
MDL编号: MFCD00000115
CAS号: 598-21-0
EINECS: 209-923-5
UN号: 2513

现在订购

产品技术规格

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5490 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR