亚油酸 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 顺-9,顺-12-十八碳二烯酸

215040050 215040250
ACRO215040050EA 0 CNY
ACRO215040050 ACRO215040250
亚油酸 99%
亚油酸
分子式: C₁₈H₃₂O₂
分子量: 280.45 g/mol
沸点: 229…230 °C (16 mmHg)
溶点: –5 °C
密度: 0.9 g/cm³
闪点: >112 °C
存储条件: Refrigerator
MDL编号: MFCD00064241
CAS号: 60-33-3
EINECS: 200-470-9
Merck Index: 13,05527

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4687 to 1.4707 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR