1,10-菲咯啉一水合物 ACS

供应商: Thermo Fisher Scientific
Danger

Synonyms: 邻菲咯啉

417120010 417120050
ACRO417120010EA 0 CNY
ACRO417120010 ACRO417120050
1,10-菲咯啉一水合物 ACS
1,10-菲咯啉一水合物
分子式: C₁₂H₈N₂·1H₂O
分子量: 198.22 g/mol
溶点: 97…101 °C
存储条件: Ambient
MDL编号: MFCD00149973
CAS号: 5144-89-8
EINECS: 200-629-2
UN号: 2811
ADR: 6.1 III
Merck Index: 13,07291
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White Crystalline powder or needles (May discolor to cream on storage)
Infrared spectrum Conforms
Suitability (for Fe determination) Passes test
UV at 450 nm A: ≤0.01
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR