1,10-菲咯啉一水合物 99+%, pure
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
邻菲咯啉
130130050
130130250
ACRO130130050EA
0
CNY
ACRO130130050
ACRO130130250
1,10-菲咯啉一水合物 99+%, pure
1,10-菲咯啉一水合物
分子式:
C₁₂H₈N₂·1H₂O 分子量: 198.22 g/mol 溶点: 97…101 °C 存储条件: Ambient |
MDL编号:
MFCD00149973 CAS号: 5144-89-8 EINECS: 200-629-2 UN号: 2811 ADR: 6.1 III Merck Index: 13,07291 |
|
产品技术规格
Appearance | White to cream Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 98 to 102 °C |
Titration with HClO4 | ≥99.0 % |
UV | at 260 nm A: ≤0.02 |
Titration after Ion exchange | ≥99.0 % (on dry substance) |
Membrane filtration test | (on 0.5 M soln.) Passes test |
Suitability | (HPLC gradient) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...