(R)-(+)-α-甲基苄胺 99+%, (99% ee)
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
(R)-(+)-1-苯乙胺
151820250
151821000
ACRO151820250EA
0
CNY
ACRO151820250
ACRO151821000
(R)-(+)-α-甲基苄胺 99+%, (99% ee)
(R)-(+)-α-甲基苄胺
分子式:
C₈H₁₁N 分子量: 121.18 g/mol 沸点: 184…186 °C (760 mmHg) 溶点: –10 °C 密度: 0.95 g/cm³ 闪点: 75 °C 存储条件: Ambient |
MDL编号:
MFCD00064405 CAS号: 3886-69-9 EINECS: 223-423-4 UN号: 2922 ADR: 8 II Merck Index: 12,06106 |
产品技术规格
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5250 to 1.5270 (20°C, 589 nm) |
GC | ≥99.0 % |
Enantiomeric excess | ≥98.5 % (GC) |
Specific optical rotation |
(20°C, 589 nm) |
Titration with HClO4 | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...