(R)-(-)-2-辛醇 99+%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
(R)-(-)-2-Hydroxy octane, (R)-2-Octanol, L(-)-2-Octanol, (R)-(-)-Hexyl methyl carbinol, (R)-(-)-Methyl hexyl carbinol
164590050
164590100
ACRO164590050EA
0
CNY
ACRO164590050
ACRO164590100
(R)-(-)-2-辛醇 99+%
(R)-(-)-2-辛醇
分子式:
C₈H₁₈O 沸点: 175 °C (760 mmHg) 密度: 0,83 g/cm³ 闪点: 60 °C 存储条件: Ambient |
MDL编号:
MFCD00064284 CAS号: 5978-70-1 EINECS: 227-777-0 UN号: 1986 ADR: 3,III Merck Index: 12,06850 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4250 to 1.4270 (20°C, 589 nm) |
GC | ≥99.0 % |
Specific optical rotation | -9° to -10° (20°C, 589 nm) (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...