(S)-(+)-2-氨基-1-丙醇 98%

供应商: Thermo Fisher Scientific
Danger

Synonyms: L-丙氨醇

104450010 104450100
ACRO104450010EA 0 CNY
ACRO104450010 ACRO104450100
(S)-(+)-2-氨基-1-丙醇 98%
(S)-(+)-2-氨基-1-丙醇
分子式: C₃H₉NO
沸点: 173…176 °C (760 mmHg)
密度: 0,965 g/cm³
闪点: 62 °C
存储条件: Ambient
MDL编号: MFCD00064412
CAS号: 2749-11-3
EINECS: 220-388-7
UN号: 2735
ADR: 8,III

现在订购

产品技术规格

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4480 to 1.4510 (20°C, 589 nm)
TLC ≥97.5 %
Specific optical rotation +20° to +24° (20°C, 589 nm) (c=2, EtOH)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR