(R)-(-)-1-氨基-2-丙醇 98+%

供应商: Thermo Fisher Scientific
Danger

Synonyms: (-)-异丙醇胺

218970010 218970050
ACRO218970010EA 0 CNY
ACRO218970010 ACRO218970050
(R)-(-)-1-氨基-2-丙醇 98+%
(R)-(-)-1-氨基-2-丙醇
分子式: C₃H₉NO
沸点: 160 °C (760 mmHg)
溶点: 23…26 °C
密度: 0,954 g/cm³
闪点: 71 °C
存储条件: Ambient
MDL编号: MFCD00064428
CAS号: 2799-16-8
EINECS: 220-532-9
UN号: 3259
ADR: 8,II

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4465 to 1.4495 (20°C, 589 nm)
GC ≥98.0 %
Specific optical rotation -16° to -20° (20°C, 589 nm) (c=1.8, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR