氯化铕(III)六水合物 99.99% (trace metals basis)
供应商: Thermo Fisher Scientific
Synonyms:
Europium(III) chloride hydrate, Europium trichloride hexahydrate
193010050
193010010
ACRO193010050EA
0
CNY
ACRO193010050
ACRO193010010
氯化铕(III)六水合物 99.99% (trace metals basis)
氯化铕(III)六水合物
分子式:
Cl₃Eu·6H₂O 分子量: 366.41 g/mol 溶点: 850 °C 密度: 4.89 g/cm³ 存储条件: Ambient |
MDL编号:
MFCD00149701 CAS号: 13759-92-7 EINECS: 233-040-4 |
产品技术规格
Appearance | White Crystalline powder or crystals and/or chunks |
Titration Complexometric | 39.0 to 42.5 % (Eu) |
Total conc. of imp.as REO | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...