氯化金(III)

供应商: Thermo Fisher Scientific
Danger

Synonyms: 四氯金酸, 四氯金酸氢, 氯金酸

223620010 223620050
ACRO223620010EA 0 CNY
ACRO223620010 ACRO223620050
氯化金(III)
氯化金(III)
分子式: HAuCl₄·H₂O
分子量: 357.8 g/mol
溶点: 30 °C
存储条件: Refrigerator
MDL编号: MFCD00149903
CAS号: 27988-77-8
EINECS: 240-948-4
UN号: 3260
ADR: 8 II

现在订购

产品技术规格

Appearance Yellow to orange Powder or crystals and/or chunks
Gold (Au) ≥49 %
Insoluble matter ≤0.1 % (in diethyl ether)
Alkalies+oth.metals (SO4) ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR