1,3,5-三羟基苯 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 间苯三酚

131040250 131041000
ACRO131040250EA 0 CNY
ACRO131040250 ACRO131041000
1,3,5-三羟基苯 99%
1,3,5-三羟基苯
分子式: C₆H₆O₃·2H₂O
分子量: 162.14 g/mol
溶点: 218…221 °C
存储条件: Ambient
MDL编号: MFCD00149090
CAS号: 6099-90-7
EINECS: 203-611-2
Merck Index: 13,07413

现在订购

产品技术规格

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR