5-磺基水杨酸 ACS

供应商: Thermo Fisher Scientific
Danger

Synonyms: 2-羟基-5-磺基苯甲酸

424511000 424515000
ACRO424511000EA 0 CNY
ACRO424511000 ACRO424515000
5-磺基水杨酸 ACS
5-磺基水杨酸
分子式: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
分子量: 254.22 g/mol
溶点: 106…110 °C
闪点: 150
存储条件: Ambient
MDL编号: MFCD00149540
CAS号: 5965-83-3
EINECS: 202-555-6
UN号: 2585
ADR: 8 III
Merck Index: 13,09054
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69e00688850e885438e2fdd9a62f6d564ebf6ae8&vid=6a3840cf38254959a0062041e9a84fa6740b1288&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White Crystalline powder or crystals or granules
Infrared spectrum Conforms
Residue after ignition ≤0.1 %
Titration with NaOH 99.0 to 101.0 %
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.02 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤0.002 %
Salicylic acid ≤0.04 % (HOC6H4COOH)
Insoluble matter ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR