1-庚烷磺酸钠一水合物 用于HPLC

供应商: Thermo Fisher Scientific
Warning

411270050 411270250 411271000
ACRO411270050EA 0 CNY
ACRO411270050 ACRO411270250 ACRO411271000
1-庚烷磺酸钠一水合物 用于HPLC
1-庚烷磺酸钠一水合物
分子式: C₇H₁₅NaO₃S·1H₂O
溶点: >300 °C
存储条件: Ambient
MDL编号: MFCD00149550
CAS号: 207300-90-1
EINECS: 245-210-5

现在订购

产品技术规格

Appearance White Powder or flakes
Infrared spectrum Conforms
Loss on drying 7.4 % to 9 % (100°C, 3 hrs) (vacuum)
Solubility (10 % w/v in water) Clear colorless
UV at 260 nm A: ≤0.01
Titration after Ion exchange ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR