EDTA tetrasodium salt hydrate ≥99.5%, Ultra Pure Grade

供应商: VWR Chemicals
Danger

Synonyms: Tetrasodium ethylenediaminetetraacetate hydrate, Kestranal® 4S hydrate, Ethylenedinitrilotetra-acetic acid tetrasodium salt hydrate, Sodium 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate xhydrate, Edetic acid tetrasodium salt hydrate, Ethylenediaminetetraacetic acid tetrasodium salt hydrate

VWRC0245-1KGEA 1704 CNY
VWRC0245-1KG VWRC0245-500G
EDTA tetrasodium salt hydrate ≥99.5%, Ultra Pure Grade
EDTA tetrasodium salt hydrate
分子式: C₁₀H₁₂N₂O₈Na₄·xH₂O
分子量: 380.17 g/mol (anhy)
溶点: >300 °C
密度: 6.9 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00150025
CAS号: 13235-36-4
EINECS: 200-573-9

现在订购

产品技术规格

Heavy Metals (as Pb) ≤0.001%
Solubility (5%, Water) pass
Iron ≤0.005%
Moisture (KF) 12.4 – 19.2%
Nitrilotriacetic Acid ≤0.1%
pH (1%, Water) @25 °C 10.7 – 11.7
Purity (Dry Basis) >99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR