碳酸胍 99+%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Guanidine carbonate salt, Diguanidinium carbonate, Guanidine carbonate
120220010
120222500
ACRO120220010EA
0
CNY
ACRO120220010
ACRO120222500
碳酸胍 99+%
碳酸胍
分子式:
C₃H₁₂N₆O₃ 溶点: 267…270 °C 密度: 1,251 g/cm³ (20 °C) 存储条件: Ambient |
MDL编号:
MFCD00013029 CAS号: 593-85-1 EINECS: 209-813-7 |
产品技术规格
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥99.0 % |
HPLC | ≥97.5 % |
pH | 10.5 to 11.5 (1 % in water) |
Specific optical rotation | -9° to -14.5° (20°C, 589 nm) (c=4 in H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...