α-甲基苄胺 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: (±)-1-苯乙胺, (±)-α-甲基苄胺

126391000 126395000
ACRO126391000EA 0 CNY
ACRO126391000 ACRO126395000
α-甲基苄胺 99%
α-甲基苄胺
分子式: C₆H₅CH(NH₂)CH₃
分子量: 121.18 g/mol
沸点: 184…186 °C (1013 hPa)
溶点: –65 °C
密度: 0.94 g/cm³
闪点: 69 °C
存储条件: Ambient
MDL编号: MFCD00008069
CAS号: 618-36-0
EINECS: 210-545-8
UN号: 2735
ADR: 8 II
Merck Index: 12,06106
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7749b7c7d0770d82ae5145b1a236458393427163&vid=9ef2ef0e518b58972043eaec1de61438a07bcc37&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5270 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.001 %
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR