2-氨基-1-丁醇 97%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-Aminobutan-1-ol, (±)-2-Amino-1-butanol
B24398.AE
B24398.AP
ALFAB24398.AEEA
0
CNY
ALFAB24398.AE
ALFAB24398.AP
2-氨基-1-丁醇 97%
2-氨基-1-丁醇
Purity: 97%
CAS No.: 96-20-8
Molecular Formula: C4H11NO
Formula Weight: 89.14
Flash Point: 89°(192°F)
Refractive Index: 1.4525
Odour: Amine-like
pH: 11.1
Melting Point: -2°
Boiling Point: 176-178°
Beilstein Reference: 1098274
Merck Reference: 14,427
UN No.: 2735
MDL No.: MFCD00008095
分子式:
C₄H₁₁NO 分子量: 89.14 g/mol 沸点: 176…178 °C 溶点: –2 °C 密度: 0.944 闪点: 89 °C (192 °F) 存储条件: Ambient |
MDL编号:
MFCD00008095 CAS号: 96-20-8 EINECS: 202-488-2 UN号: 2735 ADR: 8 III Merck Index: 13,00426 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...