2-溴-2-甲基丙烷 96% stabilised

供应商: Thermo Fisher Scientific
Danger

Synonyms: 叔丁基溴, 2-甲基-2-溴丙烷

159261000 159265000
ACRO159261000EA 0 CNY
ACRO159261000 ACRO159265000
2-溴-2-甲基丙烷 96% stabilised
2-溴-2-甲基丙烷
分子式: C₄H₉Br
分子量: 137.02 g/mol
沸点: 72…74 °C (760 mmHg)
溶点: –20 °C
密度: 1.18 g/cm³
闪点: 18 °C
存储条件: Ambient
MDL编号: MFCD00000125
CAS号: 507-19-7
EINECS: 208-065-9
UN号: 2342
ADR: 3 II
Merck Index: 13,01554

现在订购

产品技术规格

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4269 to 1.4289 (20°C, 589 nm)
GC ≥95 %
Stabilizer potassium carbonate
UV at 280 nm A: ≤0.2
pH 4.0 to 6.0 (1 M in water)
Titration with HClO4 ≥98.5 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.2 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR