碘丁烷 98% stabilised
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
丁基碘
173431000
173435000
ACRO173431000EA
0
CNY
ACRO173431000
ACRO173435000
碘丁烷 98% stabilised
碘丁烷
分子式:
C₄H₉I 沸点: 130 °C (1013 hPa) 溶点: –103 °C 密度: 1,61 g/cm³ 闪点: 33 °C 存储条件: Ambient |
MDL编号:
MFCD00001098 CAS号: 542-69-8 EINECS: 208-824-4 UN号: 2929 ADR: 6.1,II Merck Index: 13,01572 |
产品技术规格
Appearance | Clear colorless to light yellow to light pink Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4975 to 1.4995 (20°C, 589 nm) |
GC | ≥97.5 % |
Stabilizer | copper |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...