庚醛 95% stabilised

供应商: Thermo Fisher Scientific
Warning

Synonyms: 水芹醛, Oenanthaldehyde, 庚醛, 七碳醛, 正庚醛

120320500 120322500
ACRO120320500EA 0 CNY
ACRO120320500 ACRO120322500
庚醛 95% stabilised
庚醛
分子式: C₇H₁₄O
沸点: 150 °C (760 mmHg)
溶点: –42 °C
密度: 0,82 g/cm³
闪点: 35 °C
存储条件: Ambient
MDL编号: MFCD00007028
CAS号: 111-71-7
EINECS: 203-898-4
UN号: 3056
ADR: 3,III
Merck Index: 13,04677

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4110 to 1.4150 (20°C, 589 nm)
GC ≥94 %
Stabilizer 100 ppm hydroquinone
Specific gravity (25°C) 0.812 to 0.822


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR