TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether), Reagent Grade

供应商: VWR Chemicals
Danger

VWRC0747-1LEA 2262.7 CNY
VWRC0747-1L VWRC0747-4L
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether), Reagent Grade
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether)

Non-ionic - containing no charge (polyoxyethylenes or alkyl glucosides). Non-ionic detergents are suited for solubilising membrane proteins without altering biological activity.

沸点: >150 °C (1013 hPa)
密度: 1,055 g/cm³ (20 °C)
闪点: >93 °C (closed cup)
存储条件: Ambient
MDL编号: MFCD00128254
CAS号: 9036-19-5
UN号: 3082
ADR: 9,III

现在订购

产品技术规格

Cloud Point (1%, Water) 22 – 28 °C
Color (APHA) <100
Index of Refraction 1.48 – 1.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR