庆大霉素, Sigma-Aldrich®

供应商: Merck
Danger


M-Clarity™ quality level = MQ200

G1914-250MG G1914-5G
SIALG1914-250MGEA 0 CNY
SIALG1914-250MG SIALG1914-5G
庆大霉素, Sigma-Aldrich®
庆大霉素
溶点: 218…237 °C
存储条件: Ambient
MDL编号: MFCD00270181
CAS号: 1405-41-0
EINECS: 215-778-9
Merck Index: 13,04403

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR