L(-)-Glucose 98%

供应商: Thermo Fisher Scientific

Synonyms: (2S,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal

241920010 241922500
ACRO241920010EA 0 CNY
ACRO241920010 ACRO241922500
L(-)-Glucose 98%
L(-)-Glucose
分子式: C₆H₁₂O₆
沸点: 376 °C (1013 hPa)
溶点: 153…156 °C
存储条件: Refrigerator
MDL编号: MFCD00148913
CAS号: 921-60-8
EINECS: 213-068-3

现在订购

产品技术规格

Appearance White Crystalline powder
Infrared spectrum Conforms
Solubility (10 % in water) Clear colorless
HPLC ≥97.5 % (ELSD)
Specific optical rotation -50° to -54° (20°C, 589 nm) (c=10, H2O, NH3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR