二甲亚砜-d6 (99.9% D) for NMR spectroscopy
供应商: CAMBRIDGE ISOTOPE LABS
Synonyms:
二甲基亚砜-d6, DMSO-d6, 氘代二甲亚砜
DLM-10-10
DLM-10-50
DLM-10-10X0.6
CAMBDLM-10-10EA
0
CNY
CAMBDLM-10-10
CAMBDLM-10-50
CAMBDLM-10-10X0.6
二甲亚砜-d6 (99.9% D) for NMR spectroscopy
二甲亚砜-d6
分子式:
D₃CS(O)CD₃ 分子量: 84.09 g/mol 沸点: 189 溶点: 20.2 密度: 1.19 g/cm³ (20 °C) 闪点: 82 °C (closed cup) 存储条件: Ambient |
MDL编号:
MFCD00002090 CAS号: 2206-27-1 EINECS: 218-617-0 |
产品技术规格
Chemical Purity | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...